Learn More
O-fluorotoluène, 99 %, Thermo Scientific Chemicals
Catalog Number p-135884
Click to view available options
Quantité:
10 g
100 g
500 g
2.5 kg
Conditionnement:
Flacon en verre
Glass bottle
95-52-3
MFCD00000322
MMZYCBHLNZVROM-UHFFFAOYSA-N
2-fluorotoluene, o-fluorotoluene, benzene, 1-fluoro-2-methyl, toluene, o-fluoro, 1-methyl-2-fluorobenzene, unii-l55xkb3k7h, 1-fluoro-2-methyl-benzene, ortho-fluorotoluene, fluorotoluene, l55xkb3k7h
7241
1-fluoro-2-methylbenzene
CC1=CC=CC=C1F
95-52-3
MFCD00000322
MMZYCBHLNZVROM-UHFFFAOYSA-N
2-fluorotoluene, o-fluorotoluene, benzene, 1-fluoro-2-methyl, toluene, o-fluoro, 1-methyl-2-fluorobenzene, unii-l55xkb3k7h, 1-fluoro-2-methyl-benzene, ortho-fluorotoluene, fluorotoluene, l55xkb3k7h
7241
1-fluoro-2-methylbenzene
CC1=CC=CC=C1F
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Nom chimique ou matériau o-Fluorotoluene
Note relative au nom 99+%
% min. CAS 98.5
% max. CAS 100.0
Couleur Colorless to Yellow
Densité 1g/mL
Spectre IR Authentic
Plage de pourcentage du dosage 99%
Conditionnement Glass bottle
Formule moléculaire C7H7F
Formule linéaire CH3C6H4F
Indice de réfraction 1.4720 to 1.4750 (20°C, 589nm)
Quantité 10 g
Beilstein 05, 290
Merck Index 15, 4212
Poids moléculaire (g/mol) 110.131
Poids de la formule 110.13
Pourcentage de pureté ≥98.5%
Forme physique Liquid