missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OLFML3 Polyclonal Antibody

Description
Recommended positive controls: A549, HCT116. Predicted reactivity: Mouse (92%), Rat (93%), Rhesus Monkey (99%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the olfactomedin-like gene family which also includes genes encoding noelin, tiarin, myocilin, amassin, optimedin, photomedin, and latrophilin. The encoded protein is a secreted extracellular matrix glycoprotein with a C-terminal olfactomedin domain that facilitates protein-protein interactions, cell adhesion, and intercellular interactions. It serves as both a scaffold protein that recruits bone morphogenetic protein 1 to its substrate chordin, and as a vascular tissue remodeler with pro-angiogenic properties. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Feb 2017].
Spécification
Spécification
| Antigène | OLFML3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.35 mg/mL |
| Conjugué | Unconjugated |
| Formule | PBS with 20% glycerol, 1% BSA and 0.025% ProClin 300; pH 7 |
| Expression | OLFML3 |
| Numéro d’ordre du gène | Q8BK62, Q9NRN5 |
| Alias de gène | 2810002E22Rik; AI115209; AW550633; HNOEL-iso; hOLF44; mONT3; OLF44; olfactomedin like 3; olfactomedin-like 3; olfactomedin-like protein 3; OLFML3; ONT3; PSEC0035; PSEC0173; PSEC0244; UNQ663/PRO1294 |
| Symboles de gène(s) | OLFML3 |
| Afficher plus |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?