Learn More
p53 Monoclonal Antibody (OTI2F7), TrueMAB™, OriGene
Code produit 17527791
Encompass_Preferred
Change view
Click to view available options
Quantité:
30 μL
100 μL
Conditionnement:
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
17527791 30 μL 30µL
17507791 100 μL -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 17527791 Fournisseur Origene Technologies Code fournisseur TA502800S
Mouse Monoclonal Antibody
p53 Monoclonal antibody specifically detects p53 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Spécification
Antigène p53
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2F7
Concentration 0.92 mg/mL
Conjugué Unconjugated
Formule PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Expression FGFR2
Numéro d’ordre du gène P21802
Alias de gène Antigen NY-CO-13; bbl; BCC7; bfy; bhy; cellular tumor antigen p53; Cys 51 Stop; EGK_08142; FLJ92943; HGNC11998; I79_002739; LFS1; Li-Fraumeni syndrome; mutant p53; mutant tumor protein 53; OTTMUSP00000006194; p44; p53; p53 cellular tumor antigen; p53 protein; p53 tumor suppressor; p53 tumor suppressor phosphoprotein; phosphoprotein p53; Tp53; tp53.L; transformation related protein 53; transformation-related protein 53; Trp248; Trp53; tumor protein 53; tumor protein p53; tumor protein p53 (Li-Fraumeni syndrome); tumor protein p53 L homeolog; tumor suppressor p53; tumor suppressor p53 phosphoprotein; tumor suppressor protein p53; tumor supressor p53; Tumour Protein p53; XELAEV_180196761mg; Xp53; Xrel3
Espèces hôtes Mouse
Immunogène Full length human recombinant protein of human FGFR2 produced in HEK293T cell.
Méthode de purification Affinity chromatography
Quantité 30 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 2263
Espèces cibles Human
Contenu et stockage -20° C, Avoid Freeze/Thaw Cycles
Forme Liquid
Isotype IgG2a