Learn More
p63 Rabbit anti-Human, CoraLite 488, Polyclonal, Proteintech
Code produit 17781249 Tous les produits Proteintech Produits
Change view
Click to view available options
Quantité:
20 μL
100 μL
Conditionnement:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
17781249 20 μL 20µL
16850566 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 17781249 Fournisseur Proteintech Code fournisseur CL4881214320UL
Rabbit Polyclonal Antibody
The p63 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human p63. This antibody recognizes human, mouse, rat antigen. The p63 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™488.
Spécification
Antigène p63
Applications Immunofluorescence, Flow Cytometry (Intracellular Staining)
Classification Polyclonal
Concentration 1200 ug/mL
Conjugué CoraLite 488
Espèces hôtes Rabbit
Quantité 20 μL
Primaire ou secondaire Primary
Identification génétique (Entrez) 8626
Espèces cibles Human, Rat, Mouse
Isotype IgG