missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCDH18 Polyclonal Antibody

Description
Peptide sequence: MHQMNAKMHF RFVFALLIVS FNHDVLGKNL KYRIYEEQRV GSVIARLSED Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 79%; Rat: 100%.
Protocadherins comprise the largest group within the cadherin family of calcium-dependent cell-cell adhesion molecules. Protocadherin 18 (PCDH18) was initially identified through screening of human and mouse ESTs using conserved cytoplasmic domain motifs. Tissue screening revealed PCDH18 broad expression in tissues such as brain, liver, heart, kidney, lung, and trachea. PCDH18 expression is also temporally and spatially regulated in the mouse embryonic brain and interacts with Disabled-1, an intracellular adapter protein involved in neuronal migration and cell positioning during mammalian brain development. PCDH18 was also found to be expressed in the developing zebrafish neural tube and central nervous system, lending support to the hypothesis that PCDH18 may play a role during brain development.
Spécification
Spécification
| Antigène | PCDH18 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugué | Unconjugated |
| Formule | PBS with 2% sucrose and 0.09% sodium azide |
| Expression | PCDH18 |
| Numéro d’ordre du gène | Q9HCL0 |
| Alias de gène | 3110038E07Rik; BB095589; DKFZP434B0923; Kiaa1562; PCDH18; PCDH68L; Protocadherin 18; protocadherin 68-like protein; protocadherin-18 |
| Symboles de gène(s) | PCDH18 |
| Afficher plus |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?