missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Perlecan Monoclonal Antibody (A76)
GREENER_CHOICE

Code produit 11594152
Change view
Click to view available options
Quantité:
400 μL
Conditionnement:
200µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
11594152 400 μL 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 11594152 Fournisseur Invitrogen™ Code fournisseur CSI0017602

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Mouse Monoclonal Antibody

CSI 001-76-02 is highly specific for perlecan. There is no evidence for cross-reactivity with other connective tissue proteins (vitronectin, fibronectin, elastin, collagen, laminin). CSI 001-76-02 cross-reacts with human thrombospondin. Other species have not been tested. CSI 001-76-02 has been successfully used in ELISA, immunohistochemistry and Western blotting techniques. CSI 001-76-02 can be used in immunoprecipitation, ELISA and immunostaining of frozen PLP-fixed sections of bovine and human tissues. CSI 001-76-02 is more sensitive than CSI 001-71-02 or CSI 001-74-02 in ELISA. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.

Perlecan is a major heparan-sulfate proteoglycan (HSPG) found within all basement membranes and cell surfaces. Because of its strategic location and ability to store and protect growth factors, perlecan has been strongly implicated in the control of tumor cell growth and metastatic behavior. Perlecan possesses angiogenic and growth-promoting attributes primarily by acting as a coreceptor for basic fibroblast growth factor (FGF-2). Suppression of perlecan causes substantial inhibition of neoplastic growth and neovascularization. Thus, perlecan is a potent inducer of neoplasm growth and angiogenesis in vivo and therapeutic interventions targeting this key modulator of tumor progression may improve neoplastic treatment.
TRUSTED_SUSTAINABILITY

Spécification

Antigène Perlecan
Applications ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone A76
Concentration 1.01 mg/mL
Conjugué Unconjugated
Formule PBS with 15mM sodium azide; pH 7.4
Expression HSPG2
Numéro d’ordre du gène P98160
Alias de gène AI852380; Basement membrane-specific heparan sulfate proteoglycan core protein; Endorepellin; endorepellin (domain V region); heparan sulfate proteoglycan 2; HSPG; Hspg2; LG3 peptide; LOC313641; Pcn; per; perlecan; perlecan (heparan sulfate proteoglycan 2); perlecan proteoglycan; PLC; PRCAN; RP11-132G19.2; SJA; SJS; SJS1
Symboles de gène(s) HSPG2
Espèces hôtes Mouse
Immunogène Lysed bovine corneal endothelial.
Méthode de purification Protein A/G
Quantité 400 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 3339, 444872
Espèces cibles Human, Bovine
Contenu et stockage 4°C, store in dark
Type de produit Antibody
Forme Liquid
Isotype IgG1 κ
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.