missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHF7 Mouse anti-Human, Clone: OTI1F7, lyophilized, TrueMAB™
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Spermatogenesis is a complex process regulated by extracellular and intracellular factors as well as cellular interactions among interstitial cells of the testis, Sertoli cells, and germ cells. In the testis, this gene is expressed in Sertoli cells but not germ cells. However, this gene is not expressed in a patient who exhibited spermatogenic arrest at the spermatocyte stage. Spermatogenic arrest is an interruption of germ cell differentiation that may result in oligospermia or azoospermia. The proteins encoded by this gene contain plant homeodomain (PHD) finger domains, also known as leukemia associated protein (LAP) domains, believed to be involved in transcriptional regulation. Thus this protein, which localizes to the nucleus of transfected cells, has been implicated in the transcriptional regulation of spermatogenesis. Two protein isoforms are encoded by transcript variants of this gene.
Spécification
Spécification
| Antigène | PHF7 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI1F7 |
| Conjugué | Unconjugated |
| Formule | PBS with 8% trehalose and no preservative; pH 7.3 |
| Expression | PHF7 |
| Numéro d’ordre du gène | Q9BWX1 |
| Alias de gène | HSPC045, HSPC226, NYD-SP6 |
| Symboles de gène(s) | PHF7 |
| Afficher plus |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?