Learn More
Invitrogen™ Phospho-ATM (Ser1981) Monoclonal Antibody (10H11)
Description
MA1-2020 detects the phospho-ATM kinase in human and mouse samples. MA1-2020 has been successfully used in immunofluorescence, immunoprecipitation and Western blot procedures. By Western blot this antibody detects a ~370 kDa protein representing the phospho-ATM kinase in crude lysates from gamma irradiated HeLa cells. In immunofluorescence procedures, MA1-2020 recognizes the phospho-ATM kinase in irradiated human and mouse fibroblasts. The MA1-2020 immunogen is a phosphorylated synthetic peptide corresponding to the residues S(1974) L A F E E S(p) Q S T T I S S(1988) of human ATM Kinase protein.
Spécification
Spécification
| Antigène | Phospho-ATM (Ser1981) |
| Applications | Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 10H11 |
| Concentration | 1 mg/mL |
| Conjugué | Unconjugated |
| Formule | PBS with 1mg/mL BSA and 0.05% sodium azide |
| Expression | ATM |
| Numéro d’ordre du gène | Q13315, Q62388 |
| Alias de gène | AI256621; AT mutated; A-T mutated; A-T mutated homolog; AT mutated protein; AT1; ATA; Ataxia t; Ataxia telangiectasia gene mutated in human beings; ataxia telangiectasia mutated; Ataxia telangiectasia mutated homolog; ATC; ATD; ATDC; ATE; Atm; ATM serine/threonine kinase; C030026E19Rik; DKFZp781A0353; EC 2.7.1.37; MGC74674; serine-protein kinase ATM; TEL1; TEL1, telomere maintenance 1, homolog; TELO1 |
| Afficher plus |
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.