missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-EpoR (Tyr456) Polyclonal Antibody

Description
Antibody detects endogenous levels of Epo-R only when phosphorylated at Tyr456.
EpoR encodes the erythropoietin receptor, which belongs to the cytokine receptor family. When erythropoietin binds to this receptor, it activates the Jak2 tyrosine kinase, which in turn activates various intracellular pathways including Ras/MAP kinase, phosphatidylinositol 3-kinase, and STAT transcription factors. The stimulated erythropoietin receptor plays a role in the survival of erythroid cells. Defects in this gene may result in erythroleukemia and familial erythrocytosis. Dysregulation of this gene may also affect the growth of certain tumors. Multiple transcript variants are produced as a result of alternate splicing. Diseases associated with EPOR include Erythrocytosis, Familial, 1 and Erythroleukemia.
Spécification
Spécification
| Antigène | Phospho-EpoR (Tyr456) |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugué | Unconjugated |
| Formule | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Expression | EPOR |
| Numéro d’ordre du gène | P14753, P19235, Q07303 |
| Alias de gène | EPO R; EPOR; EPO-R; Erythropoietin receptor; pEpoR |
| Symboles de gène(s) | EPOR |
| Afficher plus |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?