missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-PKA beta (Thr198) Polyclonal Antibody
GREENER_CHOICE

Code produit 17277383 Tous les produits Thermo Scientific Produits
Change view
Click to view available options
Quantité:
100 μL
Conditionnement:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
17277383 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 17277383 Fournisseur Invitrogen™ Code fournisseur PA5114622

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of PKA beta only when phosphorylated at Thr198, which site historically referenced as Thr197.

The catalytic subunit C-beta of PKA (PRKACB) is a member of the Ser/Thr protein kinase family (the PKA catalytic subunit consist of three gene products: C-alpha, C-beta, and C-gamma) and has been assigned to human chromosome region 1p36.1. PRKACB is derived from a gene distinct from C-alpha and shows tissue-specific expression. At the amino acid level C-alpha and C-beta showed 93% homology. The inactive holoenzyme of AMPK is a tetramer composed of two regulatory and two catalytic subunits. cAMP causes the dissociation of the inactive holoenzyme into a dimer of regulatory subunits bound to four cAMP and two free monomeric catalytic subunits. Four different regulatory subunits and three catalytic subunits of AMPK have been identified in humans.
TRUSTED_SUSTAINABILITY

Spécification

Antigène Phospho-PKA beta (Thr198)
Applications Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugué Unconjugated
Formule PBS with 50% glycerol and 0.02% sodium azide
Expression PRKACB
Numéro d’ordre du gène P22694, P68181, P68182
Alias de gène cAMP-dependent protein kinase C beta; cAMP-dependent protein kinase catalytic beta subunit isoform 4ab; cAMP-dependent protein kinase catalytic subunit beta; PKA C-beta; Pkacb; PRKACB; protein kinase A catalytic subunit beta; protein kinase cAMP-activated catalytic subunit beta; protein kinase, cAMP dependent, catalytic, beta; protein kinase, cAMP-dependent, beta catalytic subunit; protein kinase, cAMP-dependent, catalytic, beta
Symboles de gène(s) PRKACB
Espèces hôtes Rabbit
Immunogène A synthesized peptide derived from human PRKACB(Accession P22694), corresponding to amino acid residues around phosphorylated Thr198.
Méthode de purification Sequential chromatography
Quantité 100 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 18749, 293508, 5567
Espèces cibles Human, Mouse, Rat
Contenu et stockage -20°C
Type de produit Antibody
Forme Liquid
Isotype IgG
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.