missing translation for 'onlineSavingsMsg'
Learn More

Phospho-XIAP (Ser87) Rabbit Polyclonal Antibody, Proteintech

Code produit 30233424 Tous les produits Proteintech Produits
Modifier l'affichage
Click to view available options
Quantité:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité unitSize
30233424 100 μL 100µL
30234221 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30233424 Supplier Proteintech Supplier No. 287911AP100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The Phospho-XIAP (Ser87) antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Phospho-XIAP (Ser87). This antibody recognizes human, mouse, rat antigen. The Phospho-XIAP (Ser87) antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Antigène XIAP (Ser87)
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 255 μg/mL
Conjugué Unconjugated
Symboles de gène(s) API3; BIRC4; ILP1; MIHA; XLP2
Espèces hôtes Rabbit
Immunogène Peptide
Méthode de purification Antigen affinity purification
Quantité 100 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 331
Espèces cibles Human, Mouse, Rat
Contenu et stockage Store at -20°C.
Poids moléculaire des anticorps 57 kDa
Type de produit Polyclonal Antibody
Forme Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.