Learn More
MilliporeSigma™ Polishers for Milli-Q™ IQ 7000 Ultrapure Water System
Our Lab Water purification system portfolio offers a broad range of pure and ultrapure water purification systems and services designed for scientists working in pharmaceutical, clinical, academic, industrial, research, and government laboratories — in both validated and non-validated environments.
Marque: MilliporeSigma™ CDUFBI0A1
Informations supplémentaires : Poids : 0.02000kg
Description
Our Lab Water purification system portfolio offers a broad range of pure and ultrapure water purification systems and services designed for scientists working in pharmaceutical, clinical, academic, industrial, research, and government laboratories — in both validated and non-validated environments.
Ultrafilter for the production of pyrogen-, nuclease-, protease- and bacteria-free water at the point of dispense of Milli-Q™ IQ 7000 system.Spécification
Delivered hermetically sealed | |
Biopak™ Polisher | |
No |
Milli-Q IQ 7000 Ultrapure Water System | |
Ultrafilter for the production of pyrogen-, nuclease-, protease- and bacteria-free water at the point of dispense of Milli-Q IQ 7000 system |