missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ POLR3A Polyclonal Antibody
GREENER_CHOICE

Code produit 13219918
Change view
Click to view available options
Quantité:
100 μL
Conditionnement:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
13219918 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 13219918 Fournisseur Invitrogen™ Code fournisseur PA521730

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Rabbit Polyclonal Antibody

Recommended positive controls: IMR32, SK-N-AS. Predicted reactivity: Mouse (97%), Zebrafish (94%), Xenopus laevis (94%), Chicken (97%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. POLR3A is the largest and catalytic core component of RNA polymerase III which synthesizes small RNAs, such as 5S rRNA and tRNAs. It forms the polymerase active center together with the second largest subunit. A single stranded DNA template strand of the promoter is positioned within the central active site cleft of Pol III. A bridging helix emanates from RPC1 and crosses the cleft near the catalytic site and is thought to promote translocation of Pol III by acting as a ratchet that moves the RNA-DNA hybrid through the active site by switching from straight to bent conformations at each step of nucleotide addition.
TRUSTED_SUSTAINABILITY

Spécification

Antigène POLR3A
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.75 mg/mL
Conjugué Unconjugated
Formule PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Expression POLR3A
Numéro d’ordre du gène O14802
Alias de gène 9330175N20Rik; ADDH; BC053071; DNA-directed RNA polymerase III largest subunit; DNA-directed RNA polymerase III subunit A; DNA-directed RNA polymerase III subunit RPC1; HLD7; hRPC155; POLR3A; polymerase (RNA) III (DNA directed) polypeptide A; polymerase (RNA) III (DNA directed) polypeptide A, 155kDa; polymerase (RNA) III subunit A; RGD1305574; RNA polymerase III 155 kDa subunit; RNA polymerase III subunit A; RNA polymerase III subunit C1; RNA polymerase III subunit C160; RNA polymerase III subunit RPC155-D; RPC1; RPC155
Symboles de gène(s) POLR3A
Espèces hôtes Rabbit
Immunogène Recombinant protein encompassing a sequence within the N-terminus region of human POLR3A. The exact sequence is proprietary.
Méthode de purification Antigen affinity chromatography
Quantité 100 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 11128
Espèces cibles Human
Contenu et stockage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Type de produit Antibody
Forme Liquid
Isotype IgG
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.