missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acétate de potassium, puriss. p.a., Honeywell Fluka™
35.19€ - 145.17€
Identifiants chimiques
CAS | 127-08-2 |
---|---|
Formule moléculaire | C2H3KO2 |
Poids moléculaire (g/mol) | 98.142 |
Numéro MDL | MFCD00012458 |
Clé InChI | SCVFZCLFOSHCOH-UHFFFAOYSA-M |
Synonyme | potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 |
CID PubChem | 517044 |
ChEBI | CHEBI:32029 |
Nom IUPAC | potassium;acétate |
SMILES | CC(=O)[O-].[K+] |
Description
Identifiants chimiques
127-08-2 | |
98.142 | |
SCVFZCLFOSHCOH-UHFFFAOYSA-M | |
517044 | |
potassium;acétate |
C2H3KO2 | |
MFCD00012458 | |
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 | |
CHEBI:32029 | |
CC(=O)[O-].[K+] |
Spécification
Potassium Acetate | |
C2H3KO2 | |
MFCD00012458 | |
3595449 | |
SCVFZCLFOSHCOH-UHFFFAOYSA-M | |
potassium;acétate | |
517044 | |
98.14g/mol | |
puriss. p.a. |
127-08-2 | |
CH3COOK | |
NONH for all modes of transport | |
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 | |
CC(=O)[O-].[K+] | |
98.142 | |
CHEBI:32029 | |
≥99% (perchloric acid titration) | |
1.57g/cm3at 25°C |
Sécurité et traitement
missing translation for 'einecsNumber' : 204-822-2
missing translation for 'rtecsNumber' : AJ3325000