Learn More
Fisherbrand™ Prefiltration Kit (three-stage)
Includes three stages of prefiltration. Fisher Scientific™ Prefiltration Kit protects RO membranes by removing organics, chlorine, and particles from tap water prior to RO filtration.
Marque: Fisherbrand™
Code nomenclature Nacres: JA.13
Informations supplémentaires : Poids : 6.32000kg
Description
- Activated carbon
- Anti-scaling
- Depth filtration
Spécification
3-Stage, 254mm (10 in.) | |
Accu500 | |
Water purification |