missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Prostaglandin E Receptor EP1 Polyclonal Antibody
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
The protein encoded by this gene is a member of the G protein-coupled receptor family. This protein is one of four receptors identified for prostaglandin E2 (PGE2). Through a phosphatidylinositol-calcium second messenger system, G-Q proteins mediate this receptor's activity. Knockout studies in mice suggested a role of this receptor in mediating algesia and in regulation of blood pressure. Studies in mice also suggested that this gene may mediate adrenocorticotropic hormone response to bacterial endotoxin.
Spécification
Spécification
| Antigène | Prostaglandin E Receptor EP1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugué | Unconjugated |
| Formule | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Expression | Ptger1 |
| Numéro d’ordre du gène | P34995 |
| Alias de gène | 42kDa; EP1; EP1 prostanoid receptor; PGE receptor EP1 subtype; PGE receptor, EP1 subtype; PGE2 receptor EP1 subtype; prostaglandin E receptor 1; prostaglandin E receptor 1 (subtype EP1); prostaglandin E receptor 1 (subtype EP1), 42kD; prostaglandin E receptor 1 (subtype EP1), 42kDa; prostaglandin E receptor 1, subtype EP1; prostaglandin E receptor EP1 subtype; prostaglandin E2 receptor EP1 subtype; Prostanoid EP1 receptor; PTGER1; Ptgerep1 |
| Symboles de gène(s) | Ptger1 |
| Afficher plus |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?