Description
This antibody has been tested in direct-ELISA.
ORF7 is the nucleoprotein gene of porcine reproductive and respiratory syndrome virus (PRRSV). The ORF7 encodes the nucleocapsid protein (N), the most abundant viral protein in virus-infected cells.
Spécification
Spécification
| Antigène | PRRS Virus ORF7 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 2.15 mg/mL |
| Conjugué | Unconjugated |
| Formule | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Alias de gène | ORF7 |
| Espèces hôtes | Rabbit |
| Immunogène | Recombinant Porcine reproductive and respiratory syndrome virus Nucleocapsid protein (1-123aa). |
| Méthode de purification | Protein G |
| Afficher plus |