missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PSD93 Recombinant Mouse Monoclonal Antibody (R35001_95B12)
GREENER_CHOICE

Code produit 30827101
Modifier l'affichage
Click to view available options
Quantité:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
30827101 100 μL
30825533 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 30827101 Fournisseur Invitrogen™ Code fournisseur MA563061

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Mouse Recombinant Monoclonal Antibody

PSD93 Recombinant Monoclonal Antibody for IHC (P)

PSD-93, also known as chapsyn-110, is one of a family of plasma membrane-associated proteins found in synaptic junctions. PSD-93 is unique among family members in its expression in Purkinje neuron cell bodies and dendrites. PSD-93 has three ~90 amino acid repeats called PDZ domains, a single interior SH3 domain, and a carboxyl-terminal guanylate kinase homology (GuK) domain that is enzymatically inactive. It is hypothesized that PDZ-domain interactions play a role in receptor and channel clustering which contributes to neuronal plasticiyt. PSD-93 is believed to participate in the clustering of certain proteins, including NMDA receptors and shaker-type potassium channels at the synaptic membrane. There are two principal modes of interaction between PSD-93 and other proteins. NMDA receptors and shaker-type potassium channels both share C-terminal sequence homology consisting of a threonine/serine-X-valine-COOH (T/SXV) motif. Other neuronal proteins that share this motif may interact with PSD-93 by binding to its PDZ domains. Neuronal nitric oxide synthase (nNOS), which lacks the T/SXV motif but which has its own PDZ domain, has been shown to associate with PSD-93 in vitro through a pseudo-homotypic PDZ-PDZ interaction.
TRUSTED_SUSTAINABILITY

Spécification

Antigène PSD93
Applications Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone R35001_95B12
Concentration 1 mg/mL
Conjugué Unconjugated
Formule PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Expression DLG2
Numéro d’ordre du gène Q15700
Alias de gène A330103J02Rik; B230218P12Rik; B330007M19Rik; channel-associated protein of synapse-110; channel-associated protein of synapses, 110kDa; Chapsyn-1; chapsyn-110; discs large 2; discs large homolog 2; discs large MAGUK scaffold protein 2; discs, large homolog 2; discs, large homolog 2 (Drosophila); discs, large homolog 2, chapsyn-110; disks large homolog 2; DLG2; Dlgh2; Gm1197; Postsynaptic density protein PSD-93; PPP1R58; protein phosphatase 1, regulatory subunit 58; PSD93; PSD-93; synaptic density protein PSD-93
Symboles de gène(s) DLG2
Espèces hôtes Mouse
Méthode de purification Protein A
Quantité 100 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 1740
Espèces cibles Human
Contenu et stockage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Type de produit Antibody
Forme Liquid
Isotype IgG2a
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.