missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Melanocortin-4 R/MC4R (NP_005903) VersaClone cDNA
Melanocortin 4 Receptor
Marque: R&D Systems™ RDC0531
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.00970kg
Description
The Human Melanocortin-4 R/MC4R (NP_005903) VersaClone cDNA clone contains the complete ORF for Melanocortin-4 R, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Melanocortin-4 R insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Informations sur la commande
Ships at ambient temperature.
Spécification
hMC4R | |
10 μg at 0.2 μg/μL | |
1012bp | |
Human |
NP_005903 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
For Reasearch Use Only; Not for Use in Humans