missing translation for 'onlineSavingsMsg'
Learn More

DPF1, Rabbit, Polyclonal Antibody, Abnova™

Code produit 16003407
Change view
Click to view available options
Quantité:
100 μg
Conditionnement:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
16003407 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 16003407 Fournisseur Abnova Code fournisseur PAB18794.100ug

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Rabbit polyclonal antibody raised against synthetic peptide of DPF1.

Spécification

Antigène DPF1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugué Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of DPF1.
Dilution ELISA (1:1000-1:10000) Western Blot (1:200-1:1000) The optimal working dilution should be determined by the end user.
Formule In buffer containing 0.02% sodium azide
Expression DPF1
Alias de gène MGC150428/MGC150429/NEUD4/neuro-d4
Symboles de gène(s) DPF1
Espèces hôtes Rabbit
Immunogène A synthetic peptide corresponding to amino acids at C-terminus of human DPF1.
Méthode de purification Protein A purification
Quantité 100 μg
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 8193
Espèces cibles Human, Mouse, Rat
Contenu et stockage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Forme Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.