missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Horse IgG whole molecule,Polyclonal Secondary Antibody, Abnova™
Marque: Abnova PAB9266.2mg
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Spécification
| Rabbit anti-Horse IgG whole molecule | |
| Polyclonal | |
| Rabbit anti-horse IgG recognizes horse IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography. | |
| In 10mM KH2PO4, 150mM NaCl, pH 7.2 (0.09% sodium azide) | |
| Horse IgG whole molecule | |
| 2 mg | |
| Equine | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities. | |
| Secondary | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu