Learn More
Rabbit anti-Human IgG1 Fc Secondary Antibody (245), HRP, Novus Biologicals™
Code produit 30239677 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
0.1 mL
Conditionnement:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30239677 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 30239677 Fournisseur Novus Biologicals Code fournisseur NBP340578H
Rabbit Monoclonal Antibody
IgG1 Fc Monoclonal antibody specifically detects IgG1 Fc in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Spécification
Antigène IgG1 Fc
Applications ELISA
Classification Monoclonal
Clone 245
Conjugué HRP
Formule PBS
Espèces hôtes Rabbit
Immunogène This antibody was obtained from a rabbit immunized with purified, recombinant Human IgG1 Fc (Uniprot#: P01857-1; Glu99-Lys330,C103S). HEK293 used as expression system.
Méthode de purification Protein A purified
Quantité 0.1 mL
État réglementaire RUO
Primaire ou secondaire Secondary
Espèces cibles Human
Contenu et stockage Store at 4°C in the dark.
Forme Purified
Isotype IgG