Learn More
NAE1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Description
The protein encoded by this gene binds to the beta-amyloid precursor protein. Beta-amyloid precursor protein is a cell surface protein with signal-transducing properties, and it is thought to play a role in the pathogenesis of Alzheimer's disease. In addition, the encoded protein can form a heterodimer with UBE1C and bind and activate NEDD8, a ubiquitin-like protein. This protein is required for cell cycle progression through the S/M checkpoint. Three transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Spécification
Spécification
| Antigène | NAE1 |
| Applications | Immunohistochemistry (PFA fixed), Western Blot |
| Classification | Polyclonal |
| Conjugué | Unconjugated |
| Description | Rabbit polyclonal antibody raised against synthetic peptide of NAE1. |
| Dilution | Western Blot (1:1000) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. |
| Formule | In PBS (0.09% sodium azide) |
| Expression | NAE1 |
| Alias de gène | A-116A10.1/APPBP1/HPP1/ula-1 |
| Symboles de gène(s) | NAE1 |
| Afficher plus |
For Research Use Only
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.