Learn More
Invitrogen™ Rabbit anti-Pig IgG (H+L) Secondary Antibody, FITC
Rabbit Polyclonal Secondary Antibody
Marque: Invitrogen™ 619111
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Rabbit anti-Pig IgG (H+L) Secondary Antibody for Western Blot, IHC (P), IP, ELISA
Anti-Pig secondary antibodies are affinity-purified antibodies with well-characterized specificity for porcine (swine, pig) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Spécification
| Pig IgG (H+L) | |
| Polyclonal | |
| FITC | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| Pig | |
| Secondary Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 40% glycerol and 0.1% sodium azide; pH 7.4 | |
| Gamma Immunoglobin. | |
| 2 mL | |
| Secondary | |
| 4°C, store in dark | |
| Liquid |