Learn More
SLITRK6 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Description
Members of the SLITRK family, such as SLITRK6, are integral membrane proteins with 2 N-terminal leucine-rich repeat (LRR) domains similar to those of SLIT proteins (see SLIT1; MIM 603742). Most SLITRKs, including SLITRK6, also have C-terminal regions that share homology with neurotrophin receptors (see NTRK1; MIM 191315). SLITRKs are expressed predominantly in neural tissues and have neurite-modulating activity (Aruga et al., 2003 [PubMed 14557068]).[supplied by OMIM
Spécification
Spécification
| Antigène | SLITRK6 |
| Applications | Immunohistochemistry (PFA fixed), Western Blot |
| Classification | Polyclonal |
| Conjugué | Unconjugated |
| Description | Rabbit polyclonal antibody raised against synthetic peptide of SLITRK6. |
| Dilution | Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. |
| Formule | In PBS (0.02% sodium azide) |
| Expression | SLITRK6 |
| Alias de gène | MGC119595/MGC119596/MGC119597 |
| Symboles de gène(s) | SLITRK6 |
| Afficher plus |
For Research Use Only
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.