Learn More
ACROBiosystems Rabbit CD3 epsilon&CD3 gamma Heterodimer Protein, His Tag&Flag Tag (MALS verified)
Marque:  ACROBiosystems CDG-RB52D5-100ug
Informations supplémentaires : Poids : 0.00010kg
Code produit. 17266160
  • 399.17€ / 100µg
Expédition estimée: 10-01-2023
Description
T-cell surface glycoprotein CD3 delta & CD3 gamma chain, also known as CD3D & CD3G or CD3D&CD3G respectively, are single-pass type I membrane proteins. CD3D, together with CD3- epsilon(CD3E) , CD3-gamma and CD3-zeta, and the T-cell receptor alpha/beta and gamma/delta heterodimers, forms the T cell receptor-CD3 complex. T cell receptor-CD3 complex plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways.
Rabbit CD3E&CD3G Heterodimer Protein, His Tag&Flag Tag is produced by co-expression of CD3E and CD3G, has a calculated MW of 16.5 kDa (CD3E) and 15.0 kDa (CD3G). Subunit CD3E is fused with a polyhistidine tag at the C-terminus and subunit CD3G is fused wi
Spécification
Spécification
N/A/XP_002722229.1
>90% as determined by SDS-PAGE.
Rabbit CD3E&CD3G; Heterodimer Protein, His Tag&Flag; Tag (CDG-RB52D5) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Thr 120 (CD3E) & Gln 23 - Asn 111 (CD3G) (Accession # Q9TUF9-1 (CD3E) & G1SDA3-1 (CD3G)).
-20°C
CD3E & CD3G
His Tag & Flag Tag
Less than 1.0 EU / µg by the LAL method.
Powder
Rabbit
16.5 kDa/15.0 kDa
100 µg
Gln 22 - Thr 120 (CD3E) & Gln 23 - Asn 111 (CD3G)
CD3 epsilon & CD3 gamma,CD3E & CD3G
Unconjugated
PBS, pH7.4
HEK293
Bio-Markers & CD Antigens