Learn More
MedChemExpress Ramucirumab
Code produit 16492288
Encompass_Preferred
Change view
Click to view available options
Quantité:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16492288 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16492288 Fournisseur MedChemExpress Code fournisseur HYP9920
MedChemExpress Ramucirumab is a human VEGFR-2 antagonist for the treatment of solid tumors. Ramucirumab is a recombinant human immunoglobulin G1 monoclonal antibody that binds to the extracellular binding domain of VEGFR-2 and prevents the binding of VEGFR ligands: VEGF-A, VEGF-C, and VEGF-D. Ramucirumab is also an angiogenesis inhibitor.
Spécification
Antigène VEGFR
Quantité 5 mg
Contenu et stockage Please store the product under the recommended conditions in the Certificate of Analysis.
Type de produit Inhibitory Antibodies
Conditions d’expédition Room temperature
CAS 947687-13-0
Pureté ou qualité 99.4%