missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RASGRP4 Polyclonal Antibody

Description
Recommended positive controls: Jurkat. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The RASGRP4 (RAS Guanyl Releasing Protein 4) gene encodes a guanine nucleotide exchange factor (GEF) that specifically activates RAS proteins by facilitating the exchange of GDP for GTP. Located on chromosome 19, RASGRP4 is expressed primarily in hematopoietic cells, including mast cells and certain myeloid cells. The protein contains a diacylglycerol (DAG)-binding C1 domain, a Ras exchange motif, and a CDC25 domain, which are essential for its activation and function. RASGRP4 plays a pivotal role in signal transduction pathways that regulate cell proliferation, differentiation, and survival. Dysregulation of RASGRP4 has been linked to various diseases, including inflammatory disorders and cancers, particularly those involving the hematopoietic system. Studies have shown that overexpression or mutations in RASGRP4 can lead to aberrant RAS signaling, contributing to oncogenesis and disease progression.
Spécification
Spécification
| Antigène | RASGRP4 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugué | Unconjugated |
| Formule | PBS with 40% glycerol and 0.01% thimerosal; pH 7 |
| Expression | RASGRP4 |
| Numéro d’ordre du gène | Q8TDF6 |
| Alias de gène | guanyl nucleotide releasing protein 4; RAS guanyl releasing protein 4; RAS guanyl-releasing protein 4; RASGRP4 |
| Symboles de gène(s) | RASGRP4 |
| Afficher plus |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?