Description
Regulates the p53 pathway to control the expansion growth of digestive organs.
Spécification
Spécification
| Antigène | RBM16 |
| Applications | Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | 0.33 mg/mL |
| Conjugué | Unconjugated |
| Formule | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Expression | SCAF8 |
| Numéro d’ordre du gène | Q6DID3, Q9UPN6 |
| Alias de gène | CCAP7, KIAA1116, RBM16, RNA binding motif protein 16, RNA binding protein 16, SCAF8 |
| Symboles de gène(s) | Scaf8 |
| Afficher plus |