missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant B. abortus vjbR Protein
A DNA sequence encoding the Brucella abortus (strain 2308) vjbR, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP7642-ec-200ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
WP_002966460.1 | |
vjbR Protein | |
Research Use Only | |
vjbR | |
Recombinant Protein | |
E. coli | |
MSLDLVHFPN YKKTFFGSSF QSDTLALLTR IRDEIGCRYV THTYRGRVGD CTKVNSADLT VLMTLPATWV ARYSSKNYFA IDPVFQEDAP YYRNDTSAIA RDLKEDADIC PAVAELLHDA EKHGLGNLFI AVSARNPKGV AGCTVFTFEV EDEDRTQFLA RMRPRLLSLA GIIHGTVCGC KDANSVASLL TPREVDCLRW AANGKTDGEI AEILSIARWT VVTYLQNAKI KLNCSNRTSA VATALSLGII DMPEVQHLV | |
Greater than 90% as determined by SDS-PAGE. |
29595784 | |
200 μg | |
Not determined. | |
Full Length | |
B. abortus | |
His | |
Tris-based buffer, 50% glycerol |