Learn More
enQuireBio™ Recombinant Bovine CD8A / MAL Protein
A DNA sequence encoding the Bos taurus (Bovine) CD8A / MAL, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP5800-ye-500ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_776440.1 | |
CD8A / MAL Protein | |
500 μg | |
Not determined. | |
Extracellular Domain | |
Bovine | |
His | |
Tris-based buffer, 50% glycerol |
281060 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
CD8A | |
Recombinant Protein | |
Yeast | |
LSFRMSPTQK ETRLGEKVEL QCELLQSGMA TGCSWLRHIP GDDPRPTFLM YLSAQRVKLA EGLDPRHISG AKVSGTKFQL TLSSFLQEDQ GYYFCSVVSN SILYFSNFVP VFLPAKPATT PAMRPSSAAP TSAPQTRSVS PRSEVCRTSA GSAVDTSRLD FACN |