Learn More
enQuireBio™ Recombinant Cat Serum amyloid A Protein
A DNA sequence encoding the Felis catus (Cat) (Felis silvestris catus) Serum amyloid A protein, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP6645-ec-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
Cat Serum amyloid A Protein | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Rat | |
His-B2M | |
Tris-based buffer, 50% glycerol |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
SAA1 | |
Recombinant Protein | |
E. coli | |
EWYSFLGEAA QGAWDMWRAY SDMREANYIG ADKYFHARGN YDAAQRGPGG AWAAKVISDA RENSQRVTDF FRHGNSGHGA EDSKADQEWG |