Learn More
enQuireBio™ Recombinant Chicken MDH1 Protein
A cDNA sequence encoding the MDH1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12665-50000IU
Informations supplémentaires : Poids : 0.01000kg
Spécification
MDH1 Protein | |
50000 IU | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Chicken | |
Untagged | |
Each mg of protein contains 0.59 mg NaPO4. |
Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGEPIRVLVT GAAGQIAYSL LYSIAKGDVF GKEQPLVLVL LDITPMMTVL EGVVMELQDC ALPLLREVIP TDKEEVAFKD LDIAILVGSM PRREGMERKD LLKANVKIFK SQGAALDKYA KKTVKVVVVG NPANTNCLIA SKSAPSIPKE NFSCLTRLDH NRAKSQIALK LGVTSNDVKN VIIWGNHSST QYPDVNHAKV NVKGKEVGVY EAIKDDSWLK GDFILTVQQR GAAVIKARKL SSAMSAAKAI CDHVRDIWFG TPAGEFVSMG VISDGNSYGV PEDLLYSFPV VIKDKTWKFV EGLPINDFSR EKMDLTAKEL TEEKETAVEF LSSA |