missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Dengue Virus Dengue Envelope-1 Protein

A cDNA sequence encoding the Dengue Envelope-1 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP11619-1mg

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15997818

  • 6960.00€ / 1mg
Expédition estimée: 21-06-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

Dengue Envelope-1 Protein
Research Use Only
Recombinant Protein
Baculovirus
mrcvgignrd fveglsgatw vdvvlehgsc vttmakdkpt ldiellktev tnpavlrklc ieakisnttt dsrcptqgea tlveeqdanf vcrrtfvdrg wgngcglfgk gslltcakfk cvtklegkiv qyenlkysvi vtvhtgdqhq vgnettehgt iatitpqapt seiqltdyga ltldcsprtg ldfnemvllt mkekswlvhk qwfldlplpw tsgastsqet wnrqdllvtf ktahakkqea vvlgsqegam htaltgatei qtsgtttifa ghlkcrlkmd kltlkgisyv mctgsfklek evaetqhgtv lvqvkyegtd apckipfssq dekgvtqngr lvtanpivtd kekpvnieae ppfgesyivv gagekalkls wfkkg
Protein is >95% pure as determined by 12.5% SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Dengue Virus
Untagged
Dengue Envelope-1 protein solution in 1xD-PBS, pH 7.4, 0.1% Thimerosal, 5mM EDTA, 1microg/ml of Leupeptin, Aprotinin and Pepstatin A.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions