Learn More
enQuireBio™ Recombinant Dengue Virus, ST4 Dengue NS1 Protein
A cDNA sequence encoding the Dengue NS1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10497-250ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
Dengue Virus, ST4 Dengue NS1 Protein | |
250 μg | |
Untested | |
Dengue Virus, ST4 | |
Native |
>95% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
Human | |
20mM Tris-HCl pH 7.5, 300mM NaCl. |