missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Methionine aminopeptidase Protein
A DNA sequence encoding the Escherichia coli (strain K12) Methionine aminopeptidase, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP7333-ec-10ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_414710.1 | |
Methionine aminopeptidase Protein | |
Research Use Only | |
map | |
Recombinant Protein | |
E. coli | |
AISIKTPEDI EKMRVAGRLA AEVLEMIEPY VKPGVSTGEL DRICNDYIVN EQHAVSACLG YHGYPKSVCI SINEVVCHGI PDDAKLLKDG DIVNIDVTVI KDGFHGDTSK MFIVGKPTIM GERLCRITQE SLYLALRMVK PGINLREIGA AIQKFVEAEG FSVVREYCGH GIGRGFHEEP QVLHYDSRET NVVLKPGMTF TIEPMVNAGK KEIRTMKDGW TVKTKDRSLS AQYEHTIVVT DNGCEILTLR KDDTIPAIIS HDE | |
Greater than 90% as determined by SDS-PAGE. |
947882 | |
10 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |