Learn More
enQuireBio™ Recombinant E. Coli NFNB Protein
A cDNA sequence encoding the NFNB was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12838-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
NFNB Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MDIISVALKR HSTKAFDASK KLTPEQAEQI KTLLQYSPSS TNSQPWHFIV ASTEEGKARV AKSAAGNYVF NERKMLDASH VVVFCAKTAM DDVWLKLVVD QEDADGRFAT PEAKAANDKG RKFFADMHRK DLHDDAEWMA KQVYLNVGNF LLGVAALGLD AVPIEGFDAA ILDAEFGLKE KGYTSLVVVP VGHHSVEDFN ATLPKSRLPQ NITLTEV | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
E. coli | |
His | |
1 mg/ml solution containing 20mM Tris pH 8.0, 1mM DTT, 0.05M NaCl and 10% glycerol. |