Learn More
enQuireBio™ Recombinant E. Coli TNAA Protein
A cDNA sequence encoding the TNAA was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13764-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
TNAA Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
E. coli | |
His | |
The TNAA solution (0.5 mg/ml) contains Phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT. |
Greater than 95% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMENFKHL PEPFRIRVIE PVKRTTRAYR EEAIIKSGMN PFLLDSEDVF IDLLTDSGTG AVTQSMQAAM MRGDEAYSGS RSYYALAESV KNIFGYQYTI PTHQGRGAEQ IYIPVLIKKR EQEKGLDRSK MVAFSNYFFD TTQGHSQING CTVRNVYIKE AFDTGVRYDF KGNFDLEGLE RGIEEVGPNN VPYIVATITS NSAGGQPVSL ANLKAMYSIA KKYDIPVVMD SARFAENAYF IKQREAEYKD WTIEQITRET YKYADMLAMS AKKDAMVPMG GLLCMKDDSF FDVYTECRTL CVVQEGFPTY GGLEGGAMER LAVGLYDGMN LDWLAYRIAQ VQYLVDGLEE IGVVCQQAGG HAAFVDAGKL LPHIPADQFP AQALACELYK VAGIRAVEIG SFLLGRDPKT GKQLPCPAEL LRLTIPRATY TQTHMDFIIE AFKHVKENAA NIKGLTFTYE PKVLRHFTAK LKEV |