Learn More
enQuireBio™ Recombinant EBV EBNA1 Mosaic Protein
A cDNA sequence encoding the EBV EBNA1 Mosaic was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11732-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
EBNA1 Mosaic Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
EBV | |
His |
Protein is >95% pure as determined by SDS-PAGE (coomassie staining). | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
10mM PBS pH 7.6 and 10mM NaCl. |