Learn More
enQuireBio™ Recombinant EBV p23 Protein
A cDNA sequence encoding the EBV p23 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11735-1mg
Detalles adicionales : Poids : 0.01000kg
Especificaciones
p23 Protein | |
Research Use Only | |
Recombinant Protein | |
Epstein-Barr virus (EBV) | |
25mM glycine pH 9.6 and 50% glycerol. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
EBV | |
Untagged | |
EBV- p23 protein is >95% pure as determined by 10% PAGE (coomassie staining). |