Learn More
enQuireBio™ Recombinant Hepatitis B HBsAg adw2 Protein
A cDNA sequence encoding the HBsAg adw2 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12126-50ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
Hepatitis B HBsAg adw2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Sterile filtered solution containing 50mM arginine and 1M urea. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis B | |
His | |
Greater than 90.0% as determined by SDS-PAGE. |