Learn More
enQuireBio™ Recombinant HIV-2 Protease Protein
A cDNA sequence encoding the HIV-2 Protease was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12271-2ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
HIV-2 Protease Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
20mM HEPES, pH7.0 and 10% Glycerol. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
Untagged |