Learn More
enQuireBio™ Recombinant HIV Type-O Envelope Protein
Recombinant Protein
Marque: enQuireBio™ QP12237-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
HIV Type-O Envelope Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
Untagged |
Greater than 95.0% as determined by HPLC analysis and SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
1 mg/ml in water. |