missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ACADM Protein
A DNA sequence encoding the Homo sapiens (Human) ACADM, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP5605-ec-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_000007.1 | |
ACADM Protein | |
Research Use Only | |
ACADM | |
Recombinant Protein | |
E. coli | |
KANRQREPGL GFSFEFTEQQ KEFQATARKF AREEIIPVAA EYDKTGEYPV PLIRRAWELG LMNTHIPENC GGLGLGTFDA CLISEELAYG CTGVQTAIEG NSLGQMPIII AGNDQQKKKY LGRMTEEPLM CAYCVTEPGA GSDVAGIKTK AEKKGDEYII NGQKMWITNG GKANWYFLLA RSDPDPKAPA NKAFTGFIVE ADTPGIQIGR KELNMGQRCS DTRGIVFEDV KVPKENVLIG DGAGFKVAMG AFDKTRPVVA AGAVGLAQRA LDEATKYALE RKTFGKLLVE HQAISFMLAE MAMKVELARM SYQRAAWEVD SGRRNTYYAS IAKAFAGDIA NQLATDAVQI LGGNGFNTEY PVEKLMRDAK IYQIYEGTSQ IQRLIVAREH IDKYKN | |
Greater than 90% as determined by SDS-PAGE. |
34 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |