Learn More
enQuireBio™ Recombinant Human ADAM12 Protein
A cDNA sequence encoding the ADAM12 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10962-2ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
ADAM12 Protein | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The ADAM12 solution contains 25mM Sodium Acetate pH 4.8 and 50% Glycerol. |
Greater than 95.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MAARPLPVSP ARALLLALAG ALLAPCEARG VSLWNQGRAD EVVSASVGSG DLWIPVKSFD SKNHPEVLNI RLQRESKELI INLERNEGLI ASSFTETHYL QDGTDVSLAR NYTVILGHCY YHGHVRGYSD SAVSLSTCSG LRGLIVFENE SYVLEPMKSA TNRYKLFPAK KLKSVRGSCG SHHNTPNLAA KNVFPPPSQT WARRHKRETL KATKYVELVI VADNREFQRQ GKDLEKVKQR LIEIANHVDK FYRPLNIRIV LVGVEVWNDM DKCSVSQDPF TSLHEFLDWR KMKLLPRKSH DNAQLVSGVY FQGTTIGMAP IMSMCTADQS GGIVMDHSDN PLGAAVTLAH ELGHNFGMNH DTLDRGCSCQ MAVEKGGCIM NASTGYPFPM VFSSCSRKDL ETSLEKGMGV CLFNLPEVRE SFGGQKCGNR FVEEGEECDC GEPEECMNRC CNATTCTLKP DAVCAHGLCC EDCQLKPAGT ACRDSSNSCD LPEFCTGASP HCPANVYLHD GHSCQDVDGY CYNGICQTHE QQCVTLWGPG AKPAPGICFE RVNSAGDPYG NCGKVSKSSF AKCEMRDAKC GKIQCQGGAS RPVIGTNAVS IETNIPLQQG GRILCRGTHV YLGDDMPDPG LVLAGTKCAD GKICLNRQCQ NISVFGVHEC AMQCHGRGVC NNRKNCHCEA HWAPPFCDKF GFGGSTDSGP IRQADNQGLT IGILVTILCL LAAGFVVYLK RKTLIRLLFT NKKTTIEKLR CVRPSRPPRG FQPCQAHLGH LGKGLMRKPP DSYPPKDNPR RLLQCQNVDI SRPLNGLNVP QPQSTQRVLP PLHRAPRAPS VPARPLPAKP ALRQAQGTCK PNPPQKPLPA DPLARTTRLT HALARTPGQW ETGLRLAPLR PAPQYPHQVP RSTHTAYIK |