Learn More
enQuireBio™ Recombinant Human AIDA Protein
A cDNA sequence encoding the AIDA was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10986-100ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
64853 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
AIDA | |
Human | |
His | |
AIDA protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 0.2M NaCl and 1mM DTT. |
AIDA Protein | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSEVTRS LLQRWGASFR RGADFDSWGQ LVEAIDEYQI LARHLQKEAQ AQHNNSEFTE EQKKTIGKIA TCLELRSAAL QSTQSQEEFK LEDLKKLEPI LKNILTYNKE FPFDVQPVPL RRILAPGEEE NLEFEEDEEE GGAGAGSPDS FPARVPGTLL PRLPSEPGMT LLTIRIEKIG LKDAGQCIDP YITVSVKDLN GIDLTPVQDT PVASRKEDTY VHFNVDIELQ KHVEKLTKGA AIFFEFKHYK PKKRFTSTKC FAFMEMDEIK PGPIVIELYK KPTDFKRKKL QLLTKKPLYL HLHQTLHKE |