Learn More
enQuireBio™ Recombinant Human AMN Protein
A cDNA sequence encoding the AMN was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11013-50ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
AMN Protein | |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
AMN protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 10% glycerol and 1mM DTT. |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSVSKLWVP NTDFDVAANW SQNRTPCAGG AVEFPADKMV SVLVQEGHAV SDMLLPLDGE LVLASGAGFG VSDVGSHLDC GAGEPAVFRD SDRFSWHDPH LWRSGDEAPG LFFVDAERVP CRHDDVFFPP SASFRVGLGP GASPVRVRSI SALGRTFTRD EDLAVFLASR AGRLRFHGPG ALSVGPEDCA DPSGCVCGNA EAQPWICAAL LQPLGGRCPQ AACHSALRPQ GQCCDLCGAV VLLTHGPAFD LERYRARILD TFLGLPQYHG LQVAVSKVPR SSRLREADTE IQVVLVENGP ETGGAGRLAR ALLADVAENG EALGVLEATM RESGAHVWGS S |