Learn More
enQuireBio™ Recombinant Human AMT Protein
A cDNA sequence encoding the AMT was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11015-20ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
AMT Protein | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
AMT protein solution (1 mg/ml) in Phosphate Buffered Saline, 30% glycerol and 1mM DTT. |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSAQEVLRR TPLYDFHLAH GGKMVAFAGW SLPVQYRDSH TDSHLHTRQH CSLFDVSHML QTKILGSDRV KLMESLVVGD IAELRPNQGT LSLFTNEAGG ILDDLIVTNT SEGHLYVVSN AGCWEKDLAL MQDKVRELQN QGRDVGLEVL DNALLALQGP TAAQVLQAGV ADDLRKLPFM TSAVMEVFGV SGCRVTRCGY TGEDGVEISV PVAGAVHLAT AILKNPEVKL AGLAARDSLR LEAGLCLYGN DIDEHTTPVE GSLSWTLGKR RRAAMDFPGA KVIVPQLKGR VQRRRVGLMC EGAPMRAHSP ILNMEGTKIG TVTSGCPSPS LKKNVAMGYV PCEYSRPGTM LLVEVRRKQQ MAVVSKMPFV PTNYYTLK |