missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Ang K1-3 Protein

A cDNA sequence encoding the Ang K1-3 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP10520-1mg

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15994118

  • 1500.00€ / 1mg
Expédition estimée: 04-08-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

Ang K1-3 Protein
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
VYLSECKTGN GKNYRGTMSK TKNGITCQKW SSTSPHRPRF SPATHPSEGL EENYCRNPDN DPQGPWCYTT DPEKRYDYCD ILECEEECMH CSGENYDGKI SKTMSGLECQ AWDSQSPHAH GYIPSKFPNK NLKKNYCRNP DRELRPWCFT TDPNKRWELC DIPRCTTPPP SSGPTYQCLKGTGENYRGNV AVTVSGHTCQ HWSAQTPHTH NRTPENFPCK NLDENYCRNP DGKRAPWCHT TNSQVRWEYC KIPSCDSSP
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
The activity is assayed on anti-proliferation and anti-migration of endothelial cells in vitro and anti-angiogenesis in vivo. The specific activity of anti-migration of endothelial cells in vitro is 55,000 Units/mg.
Human
Untagged
Lyophilized from a 0.2?m filtered concentrated (1.0 mg/ml) solution in 20mM NaAc, pH5.5, 4% mannitol.
Vidéos
FDS
Documents
Certificats
Promotions

Promotions