Learn More
enQuireBio™ Recombinant Human ANXA7 Protein
A cDNA sequence encoding the ANXA7 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11036-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
ANXA7 Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
ANXA7 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 150mM NaCl, 1mM DTT and 40% glycerol. |
Greater than 85.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMSYPGY PPTGYPPFPG YPPAGQESSF PPSGQYPYPS GFPPMGGGAY PQVPSSGYPG AGGYPAPGGY PAPGGYPGAP QPGGAPSYPG VPPGQGFGVP PGGAGFSGYP QPPSQSYGGG PAQVPLPGGF PGGQMPSQYP GGQPTYPSQP ATVTQVTQGT IRPAANFDAI RDAEILRKAM KGFGTDEQAI VDVVANRSND QRQKIKAAFK TSYGKDLIKD LKSELSGNME ELILALFMPP TYYDAWSLRK AMQGAGTQER VLIEILCTRT NQEIREIVRC YQSEFGRDLE KDIRSDTSGH FERLLVSMCQ GNRDENQSIN HQMAQEDAQR LYQAGEGRLG TDESCFNMIL ATRSFPQLRA TMEAYSRMAN RDLLSSVSRE FSGYVESGLK TILQCALNRP AFFAERLYYA MKGAGTDDST LVRIVVTRSE IDLVQIKQMF AQMYQKTLGT MIAGDTSGDY RRLLLAIVGQ |